Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Dimethyl 4-hydroxypyridine-2,6-dicarboxylate 98%

Supplier: AMBEED, INC
Warning

A288214-1G A288214-250MG A288214-5G A288214-25G
76875-404EA 43 USD InStock
76875-404 76875-406 76875-408 77672-484
Dimethyl 4-hydroxypyridine-2,6-dicarboxylate 98%
Dimethyl 4-hydroxypyridine-2,6-dicarboxylate
Precaución: for Research Use Only


Formula: C₉H₉NO₅
MDL Number: MFCD00185834
CAS Number: 19872-91-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty