Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-methoxybenzeneboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A291657-1G A291657-250MG A291657-5G
76875-468EA 42 USD InStock
76875-468 76875-470 76875-472
3-Fluoro-5-methoxybenzeneboronic acid pinacol ester 98%
3-Fluoro-5-methoxybenzeneboronic acid pinacol ester


Formula: C₁₃H₁₈BFO₃
MDL Number: MFCD16996298
CAS Number: 1416367-00-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty