Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Potassium (3,5-dimethylisoxazol-4-yl)trifluoroborate 97%

Supplier: AMBEED, INC
Warning

A307446-1G A307446-25G A307446-5G
76875-716EA 28 USD InStock
76875-716 76875-718 76875-720
Potassium (3,5-dimethylisoxazol-4-yl)trifluoroborate 97%
Potassium (3,5-dimethylisoxazol-4-yl)trifluoroborate
Precaución: for Research Use Only


Formula: C₅H₆BF₃KNO
MDL Number: MFCD09993005
CAS Number: 1111732-84-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty