Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 6-amino-3-chloropicolinate 95%

Supplier: AMBEED, INC
Warning

A311787-100MG A311787-1G A311787-250MG A311787-5G
76875-760EA 12 USD InStock
76875-760 76875-762 76875-764 76875-766
Methyl 6-amino-3-chloropicolinate 95%
Methyl 6-amino-3-chloropicolinate
Precaución: for Research Use Only


Formula: C₇H₇ClN₂O₂
MDL Number: MFCD18257959
CAS Number: 1256835-20-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty