Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Difluoro-2-hydroxyphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A319151-100MG A319151-1G A319151-250MG
76875-822EA 25 USD InStock
76875-822 76875-824 76875-826
3,5-Difluoro-2-hydroxyphenylboronic acid 98%
3,5-Difluoro-4-hydroxyphenylboronic acid
Precaución: for Research Use Only


Formula: C₆H₅BF₂O₃
MW: 173.91 g/mol
MDL Number: MFCD03095356
CAS Number: 1150114-51-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty