Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-[N-(2-chloro-4-pyrimidinyl)methylamino]-2,3-dimethyl-2H-indazole 97%

Supplier: AMBEED, INC
Warning

A331341-10G A331341-1G A331341-5G
76876-010EA 140 USD InStock
76876-010 76876-012 76876-014
6-[N-(2-chloro-4-pyrimidinyl)methylamino]-2,3-dimethyl-2H-indazole 97%
6-[N-(2-chloro-4-pyrimidinyl)methylamino]-2,3-dimethyl-2H-indazole
Precaución: for Research Use Only


Formula: C₁₄H₁₄ClN₅
MDL Number: MFCD12923006
CAS Number: 444731-75-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty