Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Diamino-4,6-dihydroxypyrimidine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A335374-1G A335374-25G A335374-5G
76876-070EA 11 USD InStock
76876-070 76876-072 76876-074
2,5-Diamino-4,6-dihydroxypyrimidine hydrochloride 98%
2,5-Diamino-4,6-dihydroxypyrimidine hydrochloride
Precaución: for Research Use Only


Formula: C₄H₆N₄O₂
MDL Number: MFCD00082893
CAS Number: 56830-58-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty