Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Fmoc-L-homoarginine 95%

Supplier: AMBEED, INC
Warning

A339086-1G A339086-25G
76876-148EA 10 USD InStock
76876-148 76876-150
N-Fmoc-L-homoarginine 95%
N-Fmoc-L-homoarginine


Formula: C₂₂H₂₆N₄O₄
MDL Number: MFCD01861360
CAS Number: 776277-76-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty