Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Nitro-3,4-dihydronaphthalen-1(2H)-one 95%

Supplier: AMBEED, INC
Warning

A347124-1G A347124-250MG A347124-5G
76876-238EA 30 USD InStock
76876-238 76876-240 76876-242
5-Nitro-3,4-dihydronaphthalen-1(2H)-one 95%
5-Nitro-3,4-dihydronaphthalen-1(2H)-one
Precaución: for Research Use Only


Formula: C₁₀H₉NO₃
MDL Number: MFCD09751533
CAS Number: 51114-73-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty