Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(2,6-Difluorophenyl)ethylamine 95%

Supplier: AMBEED, INC
Danger

A350737-1G A350737-250MG A350737-25G A350737-5G
76876-296EA 28 USD InStock
76876-296 76876-298 76876-300 76876-302
1-(2,6-Difluorophenyl)ethylamine 95%
1-(2,6-Difluorophenyl)ethylamine
Precaución: for Research Use Only


Formula: C₈H₉F₂N
MDL Number: MFCD09041977
CAS Number: 870849-40-4
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty