Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-tert-Butyl 6-methyl 1H-indazole-1,6-dicarboxylate 95%

Supplier: AMBEED, INC
Warning

A364707-100MG A364707-250MG A364707-5G
76876-424EA 42 USD InStock
76876-424 76876-426 76876-428
1-tert-Butyl 6-methyl 1H-indazole-1,6-dicarboxylate 95%
1-tert-Butyl 6-methyl 1H-indazole-1,6-dicarboxylate
Precaución: for Research Use Only


Formula: C₁₄H₁₆N₂O₄
MDL Number: MFCD17016051
CAS Number: 1126424-50-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty