Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4-Dibromo-2,6-di-tert-butylcyclohexa-2,5-dienone 95%

Supplier: AMBEED, INC
Warning

A365641-100MG A365641-1G A365641-250MG
76876-430EA 24 USD InStock
76876-430 76876-432 76876-434
4,4-Dibromo-2,6-di-tert-butylcyclohexa-2,5-dienone 95%
4,4-Dibromo-2,6-di-tert-butylcyclohexa-2,5-dienone
Precaución: for Research Use Only


Formula: C₁₄H₂₀Br₂O
MDL Number: MFCD00856954
CAS Number: 1144-36-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty