Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

12-Bromolauric acid 97%

Supplier: AMBEED, INC
Warning

A369841-100MG A369841-1G A369841-250MG A369841-5G A369841-25G
76876-480EA 13 USD InStock
76876-480 76876-482 76876-484 77613-098 77672-844
12-Bromolauric acid 97%
12-Bromolauric acid
Precaución: for Research Use Only


Formula: BrH₂C(CH₂)₁₀COOH
MW: 279.22
MDL Number: MFCD00002738
CAS Number: 73367-80-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty