Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-(Bromomethyl)-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%

Supplier: AMBEED, INC
Danger

A369869-1G A369869-250MG A369869-5G
76876-486EA 129 USD InStock
76876-486 76876-488 76876-490
2-(2-(Bromomethyl)-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 96%
2-(2-(Bromomethyl)-5-fluorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₃H₁₇BBrFO₂
MDL Number: MFCD18434453
CAS Number: 1030832-39-3
UN: 1760
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty