Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

HDMC 98%

Supplier: AMBEED, INC
Danger

A370003-1G A370003-250MG A370003-25G A370003-5G
76876-504EA 19 USD InStock
76876-504 76876-506 76876-508 76876-510
HDMC 98%
HDMC
Precaución: for Research Use Only


Formula: C₁₃H₁₇ClF₆N₅O₂P
MDL Number: MFCD16875663
CAS Number: 1082951-62-9
UN: 1325
ADR: 4.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty