Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Amino-1-naphthol-3-sulfonic acid hydrate 95%

Supplier: AMBEED, INC
Warning

A377043-100G A377043-25G A377043-500G
76876-600EA 120 USD InStock
76876-600 76876-602 76876-604
6-Amino-1-naphthol-3-sulfonic acid hydrate 95%
6-Amino-1-naphthol-3-sulfonic acid hydrate
Precaución: for Research Use Only


Formula: C₁₀H₉NO₄S
MDL Number: MFCD00003970
CAS Number: 87-02-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty