Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1H-Pyrrolo[3,2-b]pyridine-4-oxide 97%

Supplier: AMBEED, INC
Warning

A379262-1G A379262-250MG A379262-5G
76876-642EA 29 USD InStock
76876-642 76876-644 76876-646
1H-Pyrrolo[3,2-b]pyridine-4-oxide 97%
1H-Pyrrolo[3,2-b]pyridine-4-oxide
Precaución: for Research Use Only


Formula: C₇H₆N₂O
MW: 134.14 g/mol
MDL Number: MFCD12827548
CAS Number: 1116136-36-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty