Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-2-(pyrrolidin-1-yl)benzaldehyde 98%

Supplier: AMBEED, INC
Warning

A381653-100MG A381653-1G A381653-250MG A381653-5G
76876-680EA 23 USD InStock
76876-680 76876-682 76876-684 76876-686
5-Bromo-2-(pyrrolidin-1-yl)benzaldehyde 98%
5-Bromo-2-(pyrrolidin-1-yl)benzaldehyde
Precaución: for Research Use Only


Formula: C₁₁H₁₂BrNO
MDL Number: MFCD21333082
CAS Number: 742099-40-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty