Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Clevidipine 97%

Supplier: AMBEED, INC
Warning

A387256-100MG A387256-1G A387256-25MG A387256-50MG A387256-5G
76876-750EA 18 USD InStock
76876-750 76876-752 76876-754 76876-756 77672-918
Clevidipine 97%
Clevidipine
Precaución: for Research Use Only


Formula: C₂₁H₂₃Cl₂NO₆
MW: 456.32 g/mol
MDL Number: MFCD00940070
CAS Number: 167221-71-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty