Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(2-Hydroxyethyl)benzeneboronic acid hemiester 98%

Supplier: AMBEED, INC
Warning

A403116-100MG A403116-1G A403116-250MG
76876-914EA 47 USD InStock
76876-914 76876-916 76876-918
2-(2-Hydroxyethyl)benzeneboronic acid hemiester 98%
2-(2-Hydroxyethyl)benzeneboronic acid hemiester
Precaución: for Research Use Only


Formula: C₈H₉BO₂
MW: 147.968 g/mol
MDL Number: MFCD08436050
CAS Number: 19206-51-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty