Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

DL-ɑ-Amino-ε-caprolactam hydrochloride 95%

Supplier: AMBEED, INC
Warning

A403997-10G A403997-250MG
76876-920EA 51 USD InStock
76876-920 76876-922
DL-ɑ-Amino-ε-caprolactam hydrochloride 95%
DL-alpha-Amino epsilon caprolactam hydrochloride
Precaución: for Research Use Only


Formula: C₆H₁₂N₂O
MW: 164.63 g/mol
MDL Number: MFCD06798179
CAS Number: 29426-64-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty