Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

cis-3-Aminocyclopentanol hydrochloride 98%

Supplier: AMBEED, INC
Warning

A435890-100MG A435890-1G A435890-250MG
76877-316EA 15 USD InStock
76877-316 76877-318 76877-320
cis-3-Aminocyclopentanol hydrochloride 98%
cis-3-Aminocyclopentanol hydrochloride
Precaución: for Research Use Only


Formula: C₅H₁₂ClNO
MDL Number: MFCD22378847
CAS Number: 1284248-73-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty