Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5,12-Dihydroindolo[3,2-a]carbazole 97%

Supplier: AMBEED, INC
Warning

A436825-1G A436825-250MG A436825-25G A436825-5G
76877-326EA 34 USD InStock
76877-326 76877-328 76877-330 76877-332
5,12-Dihydroindolo[3,2-a]carbazole 97%
5,12-Dihydroindolo[3,2-a]carbazole
Precaución: for Research Use Only


Formula: C₁₈H₁₂N₂
MDL Number: MFCD22200776
CAS Number: 111296-91-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty