Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(1-Methyl-7-oxo-3-propyl-6,7-dihydro-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-N-(2-(1-methylpyrrolidin-2-yl)ethyl)-4-propoxybenzenesulfonamide 98%

Supplier: AMBEED, INC
Warning

A444987-10MG A444987-1MG A444987-250MG A444987-25MG A444987-5MG
76877-430EA 26 USD InStock
76877-430 76877-432 76877-434 76877-436 76877-438
3-(1-Methyl-7-oxo-3-propyl-6,7-dihydro-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-N-(2-(1-methylpyrrolidin-2-yl)ethyl)-4-propoxybenzenesulfonamide 98%
3-(1-Methyl-7-oxo-3-propyl-6,7-dihydro-1H-pyrazolo[4,3-d]pyrimidin-5-yl)-N-(2-(1-methylpyrrolidin-2-yl)ethyl)-4-propoxybenzenesulfonamide
Precaución: for Research Use Only


Formula: C₂₅H₃₆N₆O₄S
MDL Number: MFCD20275557
CAS Number: 268203-93-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty