Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyrrolo[1,2-a]pyrazin-1(2H)-one 98%

Supplier: AMBEED, INC
Warning

A446018-100MG A446018-250MG
76877-458EA 22 USD InStock
76877-458 76877-460
Pyrrolo[1,2-a]pyrazin-1(2H)-one 98%
Pyrrolo[1,2-a]pyrazin-1(2H)-one
Precaución: for Research Use Only


Formula: C₇H₆N₂O
MDL Number: MFCD09834911
CAS Number: 136927-63-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty