Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methoxycarbonyl)isonicotinic acid 97%

Supplier: AMBEED, INC
Warning

A449051-100MG A449051-1G A449051-250MG
76877-484EA 24 USD InStock
76877-484 76877-486 76877-488
2-(Methoxycarbonyl)isonicotinic acid 97%
2-(Methoxycarbonyl)isonicotinic acid
Precaución: for Research Use Only


Formula: C₈H₇NO₄
MDL Number: MFCD00661398
CAS Number: 24195-10-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty