Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Naphthyl isocyanate 99%

Supplier: AMBEED, INC
Danger

A456160-25G A456160-5G
76877-562EA 17 USD InStock
76877-562 76877-564
1-Naphthyl isocyanate 99%
1-Naphthyl isocyanate
Precaución: for Research Use Only


Formula: C₁₁H₇NO
MDL Number: MFCD00003881
CAS Number: 86-84-0
UN: 2206
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty