Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-tert-Butyl but-3-yn-2-ylcarbamate 97%

Supplier: AMBEED, INC
Warning

A457010-1G A457010-250MG A457010-5G
76877-566EA 310.85 USD InStock
76877-566 76877-568 76877-570 76877-572
(R)-tert-Butyl but-3-yn-2-ylcarbamate 97%
(R)-tert-Butyl but-3-yn-2-ylcarbamate
Precaución: for Research Use Only


Formula: C₉H₁₅NO₂
MW: 169.22 g/mol
MDL Number: MFCD18831560
CAS Number: 118080-82-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty