Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(R)-2-Hydroxybutanoic acid 95%

Supplier: AMBEED, INC
Danger

A464153-100MG A464153-250MG A464153-1G A464153-5G
76877-646EA 6 USD InStock
76877-646 76877-648 77594-462 77673-186
(R)-2-Hydroxybutanoic acid 95%
(R)-2-Hydroxybutanoic acid
Precaución: for Research Use Only


Formula: C₄H₈O₃
MDL Number: MFCD01318570
CAS Number: 20016-85-7
UN: 1759
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty