Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Cyano-3-methylphenylboronic acid 96%

Supplier: AMBEED, INC
Warning

A473046-100MG A473046-1G A473046-250MG A473046-5G
76877-764EA 9 USD InStock
76877-764 76877-766 76877-768 76877-770
4-Cyano-3-methylphenylboronic acid 96%
4-Cyano-3-methylphenylboronic acid
Precaución: for Research Use Only


Formula: C₈H₈BNO₂
MDL Number: MFCD18394419
CAS Number: 856255-58-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty