Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-Methoxy-3-(trifluoromethyl)phenyl)acetic acid 97%

Supplier: AMBEED, INC
Warning

A475350-1G A475350-250MG A475350-5G
76877-778EA 100 USD InStock
76877-778 76877-780 76877-782
(4-Methoxy-3-(trifluoromethyl)phenyl)acetic acid 97%
(4-Methoxy-3-(trifluoromethyl)phenyl)acetic acid
Precaución: for Research Use Only


Formula: C₁₀H₉F₃O₃
MW: 234.17
MDL Number: MFCD09832302
CAS Number: 1000566-45-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty