Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl bromofluoroacetate 98%

Supplier: AMBEED, INC
Danger

A479207-25G A479207-5G
76877-808EA 61 USD InStock
76877-808 76877-810
Ethyl bromofluoroacetate 98%
Ethyl bromofluoroacetate
Precaución: for Research Use Only


Formula: BrFCHCOOCH₂CH₃
MDL Number: MFCD00042095
CAS Number: 401-55-8
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty