Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Chloropropan-2-amine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A482808-100MG A482808-1G A482808-250MG A482808-50MG
76877-854EA 64 USD InStock
76877-854 76877-856 76877-858 76877-860
1-Chloropropan-2-amine hydrochloride 95%
1-Chloropropan-2-amine hydrochloride


Formula: C₃H₉Cl₂N
MDL Number: MFCD26407822
CAS Number: 5968-21-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty