Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Chloro-2-(hydroxymethyl)pyrimidine 98%

Supplier: AMBEED, INC
Warning

A483953-100MG A483953-1G A483953-250MG
76877-876EA 47 USD InStock
76877-876 76877-878 76877-880
5-Chloro-2-(hydroxymethyl)pyrimidine 98%
5-Chloro-2-(hydroxymethyl)pyrimidine
Precaución: for Research Use Only


Formula: C₅H₅ClN₂O
MDL Number: MFCD10697136
CAS Number: 944902-98-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty