Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2,6-dichloro-4-cyanobenzoate 98%

Supplier: AMBEED, INC
Warning

A487149-100MG A487149-1G A487149-250MG
76877-930EA 21 USD InStock
76877-930 76877-932 76877-934
Methyl 2,6-dichloro-4-cyanobenzoate 98%
Methyl 2,6-dichloro-4-cyanobenzoate
Precaución: for Research Use Only


Formula: C₉H₅Cl₂NO₂
MDL Number: MFCD13194678
CAS Number: 409127-32-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty