Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(1S,8aR)-1-Isopropyl-4,7-dimethyl-1,2,3,5,6,8a-hexahydronaphthalene 95%

Supplier: AMBEED, INC
Danger

A493694-100MG A493694-1G A493694-250MG
76877-990EA 64 USD InStock
76877-990 76877-992 76877-994
(1S,8aR)-1-Isopropyl-4,7-dimethyl-1,2,3,5,6,8a-hexahydronaphthalene 95%
(1S,8aR)-1-Isopropyl-4,7-dimethyl-1,2,3,5,6,8a-hexahydronaphthalene
Precaución: for Research Use Only


Formula: C₁₅H₂₄
MDL Number: MFCD02683434
CAS Number: 483-76-1
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty