Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,4-Diaminobenzimidamide hydrochloride 98% (by HPLC)

Supplier: AMBEED, INC
Warning

A495771-100MG A495771-1G A495771-250MG
76878-008EA 79 USD InStock
76878-008 76878-010 76878-012
3,4-Diaminobenzimidamide hydrochloride 98% (by HPLC)
3,4-Diaminobenzimidamide hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₁ClN₄
MW: 186.64 g/mol
MDL Number: MFCD26142241
CAS Number: 66717-58-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty