Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Boc-D-Ser(tBu)-OH 95%

Supplier: AMBEED, INC
Warning

A500593-1G A500593-10G A500593-25G A500593-5G
76878-038EA 6 USD InStock
76878-038 77673-308 77673-310 77673-312
Boc-D-Ser(tBu)-OH 95%
Boc-D-Ser(tBu)-OH
Precaución: for Research Use Only


Formula: C₁₂H₂₃NO₅
MDL Number: MFCD09751020
CAS Number: 248921-66-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty