Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,4,5,5-Tetramethyl-2-(naphthalen-2-ylmethyl)-1,3,2-dioxaborolane 97%

Supplier: AMBEED, INC
Warning

A501665-1G A501665-250MG A501665-5G
76878-042EA 210 USD InStock
76878-042 76878-044 76878-046
4,4,5,5-Tetramethyl-2-(naphthalen-2-ylmethyl)-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-2-(naphthalen-2-ylmethyl)-1,3,2-dioxaborolane
Precaución: for Research Use Only


Formula: C₁₇H₂₁BO₂
MDL Number: MFCD23135870
CAS Number: 1379610-55-5
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty