Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Azaspiro[2.5]octane hydrochloride 98%

Supplier: AMBEED, INC
Warning

A502471-1G A502471-250MG A502471-25G A502471-5G
76878-050EA 19 USD InStock
76878-050 76878-052 76878-054 76878-056
6-Azaspiro[2.5]octane hydrochloride 98%
6-Azaspiro[2.5]octane hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₃N·ClH
MDL Number: MFCD14581272
CAS Number: 1037834-62-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty