Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(4-Chlorophenyl)ethylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A513983-1G A513983-250MG A513983-25G A513983-5G
76878-182EA 45 USD InStock
76878-182 76878-184 76878-186 76878-188
2-(4-Chlorophenyl)ethylboronic acid pinacol ester 98%
2-(4-Chlorophenyl)ethylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₄H₂₀BClO₂
MDL Number: MFCD12546184
CAS Number: 444094-88-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty