Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-(3,5-Dimethoxyphenyl)ethanamine 95%

Supplier: AMBEED, INC
Danger

A514287-1G A514287-250MG A514287-5G
76878-190EA 23 USD InStock
76878-190 76878-192 76878-194
1-(3,5-Dimethoxyphenyl)ethanamine 95%
1-(3,5-Dimethoxyphenyl)ethanamine
Precaución: for Research Use Only


Formula: C₁₀H₁₅NO₂
MDL Number: MFCD16660443
CAS Number: 97294-78-5
UN: 2735
ADR: 8,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty