Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(4-(Hydroxy(phenyl)methyl)phenyl)boronic acid 95%

Supplier: AMBEED, INC
Warning

A519444-1G A519444-250MG A519444-5G
76878-216EA 189 USD InStock
76878-216 76878-218 76878-220
(4-(Hydroxy(phenyl)methyl)phenyl)boronic acid 95%
(4-(Hydroxy(phenyl)methyl)phenyl)boronic acid
Precaución: for Research Use Only


Formula: C₁₃H₁₃BO₃
MDL Number: MFCD17015768
CAS Number: 1221823-69-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty