Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 1H-tetrazole-5-carboxylate 95%

Supplier: AMBEED, INC
Danger

A520298-10G A520298-25G
76878-224EA 10 USD InStock
76878-224 76878-226
Ethyl 1H-tetrazole-5-carboxylate 95%
Ethyl 1H-tetrazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₄H₆N₄O₂
MDL Number: MFCD00243017
CAS Number: 55408-10-1
UN: 1325
ADR: 4.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty