Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(3-Chloropropylcarbamoyl)phenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A524558-1G A524558-250MG A524558-5G
76878-248EA 107 USD InStock
76878-248 76878-250 76878-252
3-(3-Chloropropylcarbamoyl)phenylboronic acid 98%
3-(3-Chloropropylcarbamoyl)phenylboronic acid
Precaución: for Research Use Only


Formula: C₁₀H₁₃BClNO₃
MDL Number: MFCD08436014
CAS Number: 874288-10-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty