Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Norbornene-2-carboxamide (mixture of isomers) 95+%

Supplier: AMBEED, INC
Warning

A535868-100G A535868-25G A535868-500G A535868-5G
76878-374EA 46 USD InStock
76878-374 76878-376 76878-378 76878-380
5-Norbornene-2-carboxamide (mixture of isomers) 95+%
5-Norbornene-2-carboxamide (mixture of isomers)
Precaución: for Research Use Only


Formula: C₈H₁₁NO
MW: 137.18 g/mol
MDL Number: MFCD00187896
CAS Number: 95-17-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty