Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 2-bromobenzo[d]thiazole-7-carboxylate 95%

Supplier: AMBEED, INC
Warning

A539814-100MG A539814-250MG
76878-394EA 11 USD InStock
76878-394 76878-396
Ethyl 2-bromobenzo[d]thiazole-7-carboxylate 95%
Ethyl 2-bromobenzo[d]thiazole-7-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₈BrNO₂S
MDL Number: MFCD13195423
CAS Number: 579525-09-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty