Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-cyanophenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A543086-100MG A543086-1G A543086-250MG A543086-5G
76878-416EA 39 USD InStock
76878-416 76878-418 76878-420 76878-422
3-Amino-5-cyanophenylboronic acid 98%
3-Amino-5-cyanophenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₇BN₂O₂
MDL Number: MFCD03095195
CAS Number: 913943-05-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty