Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-(2-Pyridyl)propionic acid 98%

Supplier: AMBEED, INC
Warning

A569289-10G A569289-1G A569289-5G
76878-690EA 413 USD InStock
76878-690 76878-692 76878-694
3-(2-Pyridyl)propionic acid 98%
3-(2-Pyridyl)propionic acid
Precaución: for Research Use Only


Formula: C₈H₉NO₂
MDL Number: MFCD00995159
CAS Number: 15197-75-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty