Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Rocaglamide 98%

Supplier: AMBEED, INC
Warning

A570023-1MG A570023-5MG
76878-696EA 166 USD InStock
76878-696 76878-698
Rocaglamide 98%
Rocaglamide
Precaución: for Research Use Only


Formula: C₂₉H₃₁NO₇
MW: 505.56 g/mol
MDL Number: MFCD08702699
CAS Number: 84573-16-0
Order Now

 

ORDER

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty